missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Trypsin-EDTA (0.05%), phenol red
GREENER_CHOICE

Made from trypsin powder, an irradiated mixture of proteases derived from porcine pancreas

22.08 € - 88.25 €

Specifications

Chelators EDTA
Cell Type Mammalian Cells
Classification Animal Origin
Form Liquid
Product Type Cell Culture Dissociation Reagent
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11590626
GREENER_CHOICE
View Documents
Gibco™
25300062
500 mL
88.25 €
500mL
Please to purchase this item. Need a web account? Register with us today!
11580626
GREENER_CHOICE
View Documents
Gibco™
25300054
100 mL
22.08 €
100mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Due to its digestive strength, trypsin is widely used for cell dissociation, routine cell culture passaging, and primary tissue dissociation. The trypsin concentration required for dissociation varies with cell type and experimental requirements.

  • Documented traceability
  • Dual-site cGMP Manufacturing
  • Quality testing
  • Solutions tested for pH, osmolality, sterility, and performance
  • Raw materials are verified for e-beam irradiation and tested for endotoxin, PPV, PCV 1/2, mycoplasma, bacterial, fungal, and viral contamination, as well as multiple activity assays, ash analysis, and moisture analysis
  • Documented traceability available including lot traceability, animal origin certificates, lot analyses, irradiation certificates, viral inactivation summary, and supply chain transparency
Specifications

Specifications

EDTA
Animal Origin
Cell Culture Dissociation Reagent
Phenol Red, EDTA
270 - 310 mOsm/kg
Animal Origin
Mammalian Cells
Liquid
Sterile-filtered
Sustainable packaging
Gibco™
In vitro Bioassay
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 24 Months

SDS
Documents

Documents

Product Certifications