missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Zeocin™ Selection Reagent

Selection antibiotic, formulation of phleomycin D1, a glycopeptide antibiotic produced by Streptomyces verticillus

Brand:  Gibco™ R25001

380.95 EUR valid until 2025-02-21
Use promo code "24115" to get your promotional price.



 View more versions of this product

Product Code. 10072492

  • 413.00 € / Pack of 8

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Gibco™ Zeocin™ Selection Antibiotic is a formulation of phleomycin D1, a glycopeptide antibiotic produced by Streptomyces verticillus
  • Zeocin™ Selection Antibiotic acts by binding to and cleaving DNA, leading to cell death
  • It is used for selection of of mammalian, insect, yeast, bacterial, and plant cell cultures
  • Working concentration ranges between 50–400 μg/mL depending on the cell type
  • This product is supplied at 100 mg/mL in sterile water, in 8 x 1.25 mL vials

Order Info

Shipping conditions: Ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Storage conditions: -10 to -30°C
Shipping conditions: Ice
Mammalian Cell Culture, Insect Cell Culture
Antibiotic
Eukaryotic Selection/Stable Cell Line Generation, Bacterial Selection
8 x 1.25 mL
100 mg/mL
Liquid
Sterile-filtered
Zeocin™
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • zeocin 100mg/ml

Signal Word
  • Warning

Hazard Category
  • Germ cell mutagenicity Category 2

Hazard Statement
  • H341-Suspected of causing genetic defects.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : Phleomycin from Streptomyces verticillus

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.