Learn More
Gilac™ Gilactiv™ Bedpan
Gilactiv™ Bedpan is treated with the exclusive Sanitized™ technology, which protects the bedpan's surface against viruses and bacteria.
Brand:  Gilac™ G101190
Product Code. 17938881
  • 14.90 € / Each
Explore more special offers
Description
Description
  • Suitable for hospital use.
  • Translucent, level visible, with volume graduations.
  • Rounded design and very thick.
  • Not easy to break when dropped.
  • Outstanding durability even in the intensive use cycle.
  • Product is disinfected in a bedpan washer.
  • Researched product ergonomics ensure optimum cleaning.
  • Equipment at room temperature to avoid the cold sensation experienced with stainless steel equivalents.
Specifications
Specifications
Bedpan
Polyethylene
0.7 gal.
Blue
Non-sterile
2.5 L