Learn More
Epredia™ Gill™ Hematoxylin 1, 2, and 3
With three strengths available, the Epredia™ Gill™ 1, 2, 3 Hematoxylin allows complete control of staining intensities.
Products 6
Product Code Brand Type Quantity Price Quantity & Availability  
Product Code Brand Type Quantity Price Quantity & Availability  
10624009
View Documents Promotion Details
Epredia™
72404
Gill 1 Hematoxylin 1 pt. N/A
N/A
11326740
View Documents Promotion Details
Epredia™
72411
Gill 1 Hematoxylin 1 gal. N/A
N/A
10289688
View Documents Promotion Details
Epredia™
72504
Gill 2 Hematoxylin 1 pt. N/A
N/A
10444939
View Documents Promotion Details
Epredia™
72511
Gill 2 Hematoxylin 1 gal. N/A
N/A
10580139
View Documents Promotion Details
Epredia™
72604
Gill 3 Hematoxylin 1 pt. N/A
N/A
10590139
View Documents Promotion Details
Epredia™
72611
Gill 3 Hematoxylin 1 gal. N/A
N/A
Description
Description
Gill Hematoxylins are progressive nuclear stains.
  • Gill 1 is recommended for use in cytology
  • Gill 2 has applications in histology and cytology
  • Gill 3 is used in histology
  • All are pre-mixed and ready-to-use
Videos
Safety and Handling
Safety and Handling
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • H371-May cause damage to organs.
Precautionary Statement
  • P264-Wash face, hands, and anyexposed skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P271-Use only outdoors or in a well-ventilated area.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : aluminium sulphate octadecahydrate, sodium iodate, hematoxylin, ethylene glycol,diazolidinyl urea,