Learn More
Gilmont™ Gilmont™ Falling-ball Viscometer
Gilmont Falling-ball Viscometers offer a simple, accurate, low cost, absolute method for measuring viscosity of clear fluids up to 200cp.
287.00 €
Specifications
Product Type Falling Ball Viscometer
Viscosity Range Up to 200 cp
View More Specs
Products 1
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
13075706
View Documents Promotion Details
Gilmont™
08702-10
3 This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description
Description
Gilmont™ Falling-ball Viscometers offer a simple, accurate, low cost, absolute method for measuring viscosity of clear fluids up to 200cp. Easier to use than capillary type, and less expensive than rotating/sensor electronic-based devices.
  • Unique ball-release mechanism
  • Wetted parts of glass, PTFE and Viton™ seals (stainless steel and tantalum floats optional)
  • Three range options per tube allow for better accuracy in each range
  • Absolute reading, no electronics
  • Small sample size: approx. 7mL required
  • Easily used in constant temperature bath
  • Closed system ideal for highly evaporative and hydroscopically deteriorative solutions
Recommended for:
Used in lab, R and D, process, and plant quality control
Specifications
Specifications
Falling Ball Viscometer
Up to 200 cp