Learn More
Biospec Products Mini-BeadBeater Zirconia-Silicate Beads
Mini-BeadBeater Zirconia-Silicate Beads to suit your disruption application needs.
Brand:  Biospec Products 36270-62
141.60 EUR valid until 2025-03-31
Use promo code "24119" to get your promotional price.
Product Code. 13468236
  • 228.00 € / 450g
Edge
Explore more special offers
Description
Description
  • 1 year warranty.
Specifications
Specifications
Beads
0.1 to 0.15 mm
450 g (1 lb.)
Zirconia-Silicate
Mini-Bead Beater Glass Mill
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Biospec Products Mini-BeadBeater Zirconia-Silicate Beads > Mini-BeadBeater Zirconia-Silicate Beads; Diameter: 0.1 to 0.15 mm