missing translation for 'onlineSavingsMsg'
Learn More

Gilson™ MICROMANTM E Capillary Pistons
GREENER_CHOICE

Product Code. 10192852 Shop All Gilson Products
Click to view available options
Quantity:
10 x 96 Pk.
182 Pk.
192 Pk.
200 Pk.
576 Pk.
960 Pk.
Volume (Metric):
1 to 10 μL
10 to 100 μL
100 to 1000 μL
20 to 50 μL
3 to 25 μL
50 to 250 μL
Model:
CP10
CP100
CP1000
CP1000ST
CP100ST
CP10ST
CP25
CP250
CP250ST
CP25ST
CP50
CP50ST
Format:
Box
Rack, 10 x 96
Rack, 2 x 96
Unit Size:
Pack of 182
Pack of 192
Pack of 200
Pack of 576
Pack of 960
This item is not returnable. View return policy

Product Code. 10192852

Brand: Gilson™ F148413

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MICROMANTM E Capillary Pistons provide reproducible, reliable results even with the most viscous or volatile liquids.

  • Recommended for reproducible, reliable results even with the most viscous or volatile liquids.
  • Made specifically for GilsonTM MICROMANTM E pipets.
  • Available in a wide range of volumes and designs, both in sterilized and standard versions, to suit your needs.
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 1 to 10 μL
Filtered Non-filtered
Sterility Sterile
Color Clear
Autoclavable Autoclavable
Model CP10ST
Length (English) 5.39 in.
Length (Metric) 45.1 mm
Format Rack, 2 x 96
Tip Style Positive Displacement
Packaging Box
Quantity 192 Pk.
Product Type Capillary Piston
Disposable Single-use
For Use With (Equipment) Gilson Microman
Material Polypropylene
Fit for Purpose Certified Quality
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.