Learn More
Gilson™ MICROMAN™ E Capillary Pistons
GREENER_CHOICE
MICROMAN™ E Capillary Pistons provide reproducible, reliable results even with the most viscous or volatile liquids.
Brand:  Gilson™ F148415
 View more versions of this product
Product Code. 10789324
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Recommended for reproducible, reliable results even with the most viscous or volatile liquids.
  • Made specifically for Gilson™ MICROMAN™ E pipets.
  • Available in a wide range of volumes and designs, both in sterilized and standard versions, to suit your needs.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
10 to 100 μL
Sterile
Autoclavable
5.35 in.
Box
192 Pk.
Single-use
Polypropylene
Non-filtered
Clear
CP100ST
41.8 mm
Box
Capillary Piston
Gilson Microman
Certified Quality