Learn More
Gilson™ MICROMAN™ E Capillary Pistons
GREENER_CHOICE
MICROMAN™ E Capillary Pistons provide reproducible, reliable results even with the most viscous or volatile liquids.
Brand:  Gilson™ F148560
95.12 EUR valid until 2025-12-31
Use promo code "24285" to get your promotional price.
 View more versions of this product
Product Code. 12597526
  • 130.00 € / Pack of 182
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Recommended for reproducible, reliable results even with the most viscous or volatile liquids.
  • Made specifically for Gilson™ MICROMAN™ E pipets.
  • Available in a wide range of volumes and designs, both in sterilized and standard versions, to suit your needs.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
100 to 1000 μL
Non-sterile
Autoclavable
5.31 in.
Box
182 Pk.
Single-use
Polypropylene
Non-filtered
Clear
CP1000
70.4 mm
Box
Capillary Piston
Gilson Microman
Certified Quality
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Gilson™ MICROMAN™ E Capillary Pistons > MICROMAN™ E Capillary Pistons; Model: CP1000; Quantity: Box of 182; Volume: 100 to 1000 μL