Learn More
Gilson™ MICROMAN™ E Capillary Pistons
GREENER_CHOICE
MICROMAN™ E Capillary Pistons provide reproducible, reliable results even with the most viscous or volatile liquids.
Brand:  Gilson™ F148714
325.99 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 15314785
  • 457.00 € / Pack of 576
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Recommended for reproducible, reliable results even with the most viscous or volatile liquids.
  • Made specifically for Gilson™ MICROMAN™ E pipets.
  • Available in a wide range of volumes and designs, both in sterilized and standard versions, to suit your needs.
Specifications
Specifications
50 to 250 μL
Sterile
Autoclavable
10.71 in.
Box
576 Pk.
Single-use
Polypropylene
Non-filtered
Clear
CP250ST
86.45 mm
Box
Capillary Piston
Gilson Microman
Certified Quality