missing translation for 'onlineSavingsMsg'
Learn More

Gilson™ MICROMANTM E Positive Displacement Pipette
GREENER_CHOICE

Product Code. p-4932285 Shop All Gilson Products
missing translation for 'orderingAttributeHoverText'
Volume (Metric):
1 to 10 μL
10 to 100 μL
20 to 50 μL
3 to 25 μL
50 to 250 μL
Model:
M100E
M10E
M250E
M25E
M50E
missing translation for 'unitSize'
Each
This item is not returnable. View return policy

Product Code. 15314775

missing translation for 'mfr': Gilson™ FD10001

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

The ergonomically designed MICROMANTM E Positive Displacement Pipette, along with disposable capillary piston (CP) tips, provide the highest precision when pipetting viscous, non-aqueous liquids such as oils, blood, glycerol, and more, while protecting the pipette from contamination.

MICROMANTM E positive displacement pipettes with capillary pistons offer highest precision when pipetting problem liquids such as oils, cosmetics, blood, glycerol, radioactive samples, RNA, DNA, and more.

  • Ergonomic design to increase pipetting performance.
  • Patented QuickSnap feature for Capillary Piston fitting System.
  • Volume securing button to reduce risk of accidental volume change during pipetting cycles.
  • Comfortable and easy to use with new handle shape and added finger hook.
  • Fully visible volumeter.
  • Large pushbutton for allows easy aspirate and dispense.
  • For complete and rapid pipetting of viscous, volatile, dense, hot or cold liquids.
  • Avoids leaks when pipetting volatile (high vapor pressure) liquids.
  • Avoid cross-contamination or residual sample carry-over.
  • For tips, see F148412G or F148413G.
TRUSTED_SUSTAINABILITY

Specifications

Compatible Tips CP Tips
Autoclavable Autoclavable
Color Red
Material Polypropylene
Model M10E
Number of Channels 1
Programmability Yes
Type Positive Displacement Pipette
Volume (Metric) 1 to 10 μL
For Use With (Application) For pipetting viscous, volatile, and problem liquids
Quantity 1
Length (English) 4.25 in.
Width (English) 12.48 in.
Height (English) 1.42 in.
Fit for Purpose Certified Quality
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.