Learn More
Gilson™ MICROMAN™ Positive-Displacement Pipets
Ensures accuracy and precision when pipetting problem liquids, and offers full protection against cross contamination. Gilson™ MICROMAN™ Positive-Displacement Pipets are 3 to 4 times better than an air displacement pipet when pipetting viscous or volatile liquids.
Brand:  Gilson™ F148506
 View more versions of this product
Product Code. 10541113
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Piston is located inside the tip or capillary preventing pipet contamination
  • Color-coded push-button allows for easy identification of different models
  • Dual function push-button that also ejects capillary pistons
  • Comfortable PVDF handle resistant to aggressive chemicals
  • Accurately measure high vapor pressure liquids without leakage or degradation of the tip
  • M10 and M100 models are fully autoclavable
Specifications
Specifications
C1000
M1000
Positive Displacement Pipette
Pipetting of viscous and dense liquids
Autoclavable
1
100 to 1000 μL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products