Learn More
Gilson™ PIPETMAN™ DIAMOND Reload Pack Empty Box Set
GREENER_CHOICE
PIPETMAN™ Diamond Reload Pack Empty Box Set for blister refill and reload packs. Easy to store on all benches.
Brand:  Gilson™ F171204
17.46 EUR valid until 2024-11-29
Use promo code "21683" to get your promotional price.
 View more versions of this product
Product Code. 15279386
  • 21.25 € / Pack of 2
Estimated Shipment: 27-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Hold tips of model DL10 (0.1 to 20 μL), D200 (2 to 200 μL), D1000 (100 to 1000 μL)
  • 70% less plastic
  • Three times smaller than regular tips box
  • Eco-friendly
  • Available in set of 2
Specifications
Specifications
Empty Box Reload
Autoclavable
Models DL10 (0.1 to 20μL), D200 (2 to 200μL)
Rectangular
Non-sterile
9.3 cm
5.28 in.
10 x 96
Clear
Polypropylene
Yes, Interlocking
Set of 2
3.66 in.
4.92 in.