missing translation for 'onlineSavingsMsg'
Learn More

Gilson™ PIPETMAN™ EXPERT™ Universal Fit Gel Loading Pipette Tips: Nonsterile
GREENER_CHOICE

Flat or round tip styles are available

Brand:  Gilson™ F1731182

 View more versions of this product

Product Code. 15942051

  • 469.00 € / Pack of 800
Estimated Shipment: 30-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Color-coded tip packaging corresponds to industry standards for volume color matching to pipettes
  • Features graduations at varying volumes allowing for quick-checks when aspirating samples
  • Sterile model also available

Specifications

Specifications

0.1 to 10 μL
Clear
4.572cm
Yes
Polypropylene
Non-sterile
12.75 in.
Flat, Gel Loading
Pipetman EXPERT
2μL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.