missing translation for 'onlineSavingsMsg'
Learn More

Gilson™ PIPETMAN™ Starter Kit
GREENER_CHOICE

Provides greater pipetting comfort without compromising accuracy and precision. Gilson™ PIPETMAN™ Starter Kit is an economical solution for daily pipet users with concerns for the susceptibility of Repetitive Strain Injury (RSI).

Brand:  Gilson™ F167900

696.48 EUR valid until 2025-04-13
Use promo code "24122" to get your promotional price.


Product Code. 12372003

  • 897.00 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Provides greater pipetting comfort without compromising accuracy and precision. All-in-one kit is an economical solution for daily pipet users who have concerns regarding the susceptibility of Repetitive Strain Injury (RSI.)

Kit Includes:

  • Three PIPETMAN Pipettes: P20, P200, and P1000 (equipped with stainless steel ejectors)
  • Two PIPETMAN DIAMOND Tipacks: D200 and D1000 (racks of 96 tips each)
  • One Gilson Guide to Pipetting Handbook
  • Three Plastic Tip Ejectors
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Manual Air Displacement Pipette
Pipetman
Manual Air Displacement Pipette
3 year
12.01 in.
Polypropylene
Starter Kit
P20G, P200G, P1000G pipette, plus boxes of-quality, PIPETMAN Tip D200, D1000, Stainless Steel and Plastic tip ejectors, 3 Single™ pipette holders and Gilson guide to pipetteting
14.37 in.
4.72 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gilson™ PIPETMAN™ Starter Kit > Regular Kit

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.