Learn More
Gilson™ PIPETMAN™ Neo Pipets
Fits most brands of tips. Gilson PIPETMAN™ Neo Pipets are the next generation of the PIPETMAN Classic™ with lower spring and ejection forces.
Brand: Gilson™ F144565
This item is not returnable.
View return policy
Description
- Redesigned tip ejector and ejection springs reduces tip ejection forces
- Unique clip ejector fits securely into the body of the pipette to avoid slipping over time
- PVDF handle limits the transfer of heat from hand to the pipette and is resistant to aggressive chemicals
- Stainless steel tip ejector resists corrosion and offers a smooth tip ejection stroke
- Color-coded push-button for ease of identification
- Unique tip holder allows for a secure fit of most brands of tip
- Models P2N and P10N incorporate dual position tip ejectors, allowing the use of D10 (short) and DL10 (long)
Order Info
Trade in program available, please contact your Fisher Scientific Sales representative for more details.
Specifications
±0.5, ±0.8, ±1.6μL | |
D200, D300, DF100ST, DF200ST, DF300ST | |
1 | |
Pipette | |
Ergonomic |
ISO8655 | |
P200N | |
Pipetman Neo | |
20 to 200 μL | |
Color coded push button, PVDF handle, Stainless Steel tip ejectors, universal tip holder |
Product Content Correction
Product Title
Gilson™ PIPETMAN™ Neo Pipets > Model P200N; Vol.: 20 - 200μL
Spot an opportunity for improvement?Share a Content Correction