Learn More
Gilson™ PIPETMAN™ TIPACK™ Racked Pipet Tips
GREENER_CHOICE
Engineered to be functional, convenient and robust. Gilson™ PIPETMAN™ TIPACK™ Racked Pipet Tips are available in sterilized or autoclavable packages.
Brand:  Gilson™ F171101
70.33 EUR valid until 2024-12-31
Use promo code "21921" to get your promotional price.
 View more versions of this product
Product Code. 12679581
  • 86.30 € / Pack of 960
Estimated Shipment: 06-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Optimized geometry and pure polypropylene material ensure low retention and naturally hydrophobic surface.
  • Requires low tip-mounting force.
  • Robust and designed to withstand autoclaving.
  • Sterility assurance level SAL 10-6 guaranteed, using gamma irradiation.
  • All packs are wrapped in medical device quality plastic which provides a complete shield against contamination.
  • Indicator dots used on Gilson products undergo an irreversible color change upon sterilization; yellow gamma dots turn red when products have been properly exposed.
  • TIPACKs are securely sealed with a sticker; a full product description is given on the TIPACK for traceability and easy identification.
  • Available in sterilized or autoclavable packages.
  • Manufactured in a clean room and hence no risk of contamination.
  • Full traceability from the mold to the bench guaranteed.
  • Easy identification with color-coded packaging to meet GLP requirements.
Specifications
Specifications
0.1 to 10 μL
Sterile
Autoclavable
5.71 in.
Rack
Single-use
Polypropylene
Non-filtered
Clear
D10
14.5 cm
Pipet Tips
Pipetman Tipack