Learn More
Gilson™ PIPETMAN™ Universal Reload Box
GREENER_CHOICE
Required for use of Tower Pack tips. Gilson™ PIPETMAN™ Universal Reload Box is color-coded.
Brand:  Gilson™ F167100
15.38 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 10122862
  • 23.65 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • TOWERPACK™ is an innovative, quick and easy refill system for DIAMOND™ Tips
  • Uses 60% less space on your laboratory bench and 70% less plastic
  • Entire range of DIAMOND™ Tips TOWERPACK™ fits the amazingly easy-to-use universal reload box
  • Color-coded racks differentiate tip models and indicate the compatible Gilson™ pipette models
Specifications
Specifications
Red, Green, Yellow, or Blue
960
66 mm