Learn More
GIPC1 293T Cell Overexpression Lysate (Denatured), Abnova
Brand:  Abnova H00010755-T01.100uL
Product Code. 16133126
  • 215.00 € / 100µL
Explore more special offers
Description
Description
Western Blotting
Specifications
Specifications
293T
GIPC1 293T Cell Transient Overexpression Lysate(Denatured)
36
pCMV-GIPC1 full-length
100 μL
Human
Western Blot
NM_005716
6.8
Transient overexpression cell Lysate was tested with Anti-GIPC1 antibody (H00010755-B01) by western blotting
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Show More
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
GIPC1 293T Cell Overexpression Lysate (Denatured), Abnova > 100μL