Learn More
Tocris Bioscience™ GKA 50
Glucokinase activator
Brand:  Tocris Bioscience™ 5133/10
Product Code. 18744739
  • 370.00 € / 10mg
Explore more special offers
Description
Description
GKA 50 is a potent glucokinase activator (EC50 = 33 nM). Significantly reduces plasma glucose levels in diabetic rats; stimulates insulin release from mouse islets of Langerhans.
Specifications
Specifications
>98%
GKA 50
GKA 50
10 mg
464.51