missing translation for 'onlineSavingsMsg'
Learn More
Corning™ GL45 Polypropylene Screw Caps Product Code.: 17263484
Add to basket

Corning™ GL45 Polypropylene Screw Caps
GREENER_CHOICE

Product Code. 17263484 Shop All Corning Products
Green, Storage bottle, 20/Cs.
Click to view available options
Color:
Gray
Green
Light Gray
Purple
For Use With (Equipment):
Storage bottle
No. per Case:
10/Cs.
20/Cs.
This item is not returnable. View return policy

Product Code. 17263484

Brand: Corning™ 139545LTC3

Add to basket
Add to basket
This item is not returnable. View return policy

Linerless plug seal design for efficient sealing

These are various sizes and types of screw caps for use with storage bottles.

TRUSTED_SUSTAINABILITY

Specifications

Color Green
Material Polypropylene
No. per Case 20/Cs.
No. per Pack 20/Pk.
Sterility Non-sterile
Thread Size (Metric) GL 45
Closure Color Green
For Use With (Equipment) Storage bottle
Closure Material Polypropylene
Closure Type Plug Seal
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Corning™ GL45 Polypropylene Screw Caps >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.