Learn More
BRAND™ Glass Automatic Burette - BB
Without intermediate stopcock. Includes bottle.
Brand:  BRAND™ 922523
 View more versions of this product
Product Code. 15805522
  • 313.00 € / Each
Estimated Shipment: 14-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Safety stopcock
  • Individual conformity certificate
  • TRUSTED_SUSTAINABILITY
    Specifications
    Specifications
    50 mL
    0.1 mL
    Automatic Burette
    Glass
    Bottle