Learn More
Hecht Karl™ Glass Balls
Made of clear, polished glass. Karl Hecht™ Glass Balls can be used as boiling stones, mixing beads or for distillation columns.
Brand:  Hecht Karl™ 41401002
 View more versions of this product
Product Code. 10359111
  • 29.00 € / 1kg
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
 Spherical, solid, polished glass beads for general laboratory use.
Specifications
Specifications
Ball
Glass
2 mm
1kg Pack