missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Glass Beads

Contains replacement reagents to run all Saftest™ test kits.

Brand:  MP Biomedicals™ 07DP1070

Product Code. 15506236

  • 55.25 € / Each
Estimated Shipment: 20-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • For use with SafTest analyzer
  • Used with all six SafTest test kits
  • Appropriate for all testing endpoints
  • Research area: Lipid degradation and oxidation
  • Detection method: Bichromatic photometer
  • Application area: Oil/fat degradation
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Glass
One tube (5mm sized beads)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers