Learn More
RETSCH Glass Beads for Mills
For use with Retsch MM 200 Mixer Mills.
Brand:  RETSCH 22.222.0005
 View more versions of this product
Product Code. 10782701
  • 71.75 € / 500g
Estimated Shipment: 23-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Ideal for disruption of biological cells
Specifications
Specifications
Beads
1 to 1.5 mm
500 g
Glass
Mixer Mill MM 200