Learn More
RETSCH Glass Beads for Mills
For use with Retsch MM 200 Mixer Mills.
Brand:  RETSCH 22.222.0001
 View more versions of this product
Product Code. 11726175
  • 63.95 € / Each
Estimated Shipment: 10-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Ideal for disruption of biological cells
Specifications
Specifications
Beads
0.10 to 0.25 mm
500 g
Glass
Mixer Mill MM 200