Learn More
RETSCH Glass Beads for Mills
For use with Retsch MM 200 Mixer Mills.
Brand:  RETSCH 22.222.0003
 View more versions of this product
Product Code. 11746175
  • 71.75 € / 500g
Estimated Shipment: 14-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Ideal for disruption of biological cells
Specifications
Specifications
Beads
0.50 to 0.75 mm
500 g
Glass
Mixer Mill MM 200