missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Glass Bottom for Desiccator

Glass only bottom for use with desiccator

Brand:  Fisherbrand™ 632415280100

 View more versions of this product

Product Code. 15504625

  • 86.80 € / Each
Estimated Shipment: 17-02-2025
to see stock.
Explore more special offers
Add to basket

Description

Description

  • Bottom for desiccator

Certifications

ISO 13 130, DIN 12 491

Specifications

Specifications

94 mm
I.D.: 66 mm
Desiccator (Type: 100mm)
Bottom
Glass
113 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.