Learn More
Hirschmann™ Class A Volumetric Flasks with Poly Stopper
Achieve precise standard solutions. Hirschmann™ Class A Volumetric Flasks with Poly Stopper is standardized to contain or to deliver a definite amount of liquid. Flask has silver diffusion ink with brown graduations.
Brand:  Hirschmann™ 2822152
 View more versions of this product
Product Code. 13197949
  • 42.45 € / Pack of 2
Estimated Shipment: 10-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • With polyethylene stopper
  • With amber stain graduation
  • Acc. to DIN EN ISO 1042
  • Corrosion-free and resistant to thermal shock
  • Conformity certified, with dated batch identification
Specifications
Specifications
To Contain
Polyethylene
5 mL
70 ± 5 mm
± 0.025 mL
7/16 NS Standard Ground
Autoclavable
No
A
DIN EN ISO 1042
Permanent amber markings with silver diffusion color
Borosilicate Glass (3.3)
Polyethylene Stopper
2/Pack
± 0.025 mL
Reusable
Non-sterile
Round