missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BUCHI Glass Bulb Tube Ends
140.00 € - 180.00 €
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
12810723
|
BUCHI
037101 |
5 mL |
180.00 €
Each |
Estimated Shipment: 19-02-2025
Log in to see stock. |
|||||
10477053
|
BUCHI
037104 |
20 mL |
140.00 €
Each |
Estimated Shipment: 10-01-2025
Log in to see stock. |
|||||
Specifications
Bulb Tube End | |
Glass |