Learn More
Hirschmann™ ringcaps™ Disposable Capillary Pipettes
Product Code. 11772323
Click to view available options
Volume (Metric):
1 to 5 μL
10 μL
100 μL
100 to 200 μL
20 μL
20 to 40 μL
44.7 μL
50 μL
50 to 100 μL
Unit Size:
Pack of 1000
Pack of 400
Product Code. 11772323
Brand: Hirschmann™ 9600222
Only null left
Add to basket
Add to basket
Excellent for working with small samples in the laboratory. Hirschmann Instruments™ ringcaps™ Disposable Capillary Pipettes can be utilized for a wide range of scientific and medical applications.
  • With ringmark
  • Conformity certified
  • Manufactured from durable DURAN™ glass
  • Acc. to ISO 7550, CE acc. IVD 98/79 EC
Specifications
Accuracy 0.25%
Certifications/Compliance ISO 7550, CE, IVD 98/79 EC
Material Glass
Includes Ring Mark
Volume (Metric) 100 to 200 μL
Type Capillary Pipette
Disposable Single-use