Learn More
Marienfeld Superior™ Glass Capillary Tubes for the Determination of Melting Point
Use Marienfeld Superior™ Glass Capillary Tubes for storage of small amounts of liquids and to determine melting points during laboratory experimentation.
Brand: Marienfeld Superior™ 2940206
This item is not returnable.
View return policy
Description
- With rectangular cut edges
- For single use
- 100 capillaries in a dispenser vial, 10 vials in a box
Specifications
Glass | |
1 mm | |
80 mm |
Capillaty Tubes (One End Closed) | |
For Determination Of Melting Point | |
1000 |
Product Content Correction
Product Title
Marienfeld Superior™ Glass Capillary Tubes for the Determination of Melting Point > Diameter: 1mm; One End Closed
Spot an opportunity for improvement?Share a Content Correction