missing translation for 'onlineSavingsMsg'
Learn More

Bellingham + Stanley™ Glass Center Polarimeter Tubes

Compatible with most makes of polarimeter. Bellingham + Stanley™ Glass Center Polarimeter Tubes are manufactured to high standards conforming to ICUMSA recommendations. Tube ends are precision ground in order to achieve the highest accuracy optical rotation measurement.

Brand:  Bellingham + Stanley™ 35-58

 View more versions of this product

Product Code. 11970078

  • 353.00 € / Each
Estimated Shipment: 25-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Filling tubes with standard 8mm glass in the center
  • Features a funnel shaped center fill cup for viscous samples
  • Precision ground with windows made from specially selected low-strain glass
  • Available in 100mm and 200mm lengths
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Polarimeter Tube with Funnel-shaped Center Fill Cup for Viscous Samples
Glass, Standard 8 mm
Polarimeter
200 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers