Learn More
Duran™ Glass Cones with Ridge
Rigid glass connections and the most frequently used connection type in glass apparatus.
Brand: Duran™ 286191909
111.30 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- Exact cone angles for excellent quality
- Low joint surface roughness
- Acc. to DIN 12 249
- With or without tip
- Provides tight seals
Specifications
50/pk | |
Cone with ridge | |
With Tip | |
NS 19/26 |