Learn More
Duran™ Glass Cones with Ridge
Rigid glass connections and the most frequently used connection type in glass apparatus.
Brand:  Duran™ 286192905
Product Code. 15836911
  • 128.00 € / Pack of 10
Estimated Shipment: 07-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Exact cone angles for excellent quality
  • Low joint surface roughness
  • Acc. to DIN 12 249
  • With or without tip
  • Provides tight seals
TRUSTED_SUSTAINABILITY
Specifications
Specifications
50/pk
Cone with ridge
With Tip
NS 29/32