missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Glass Conical Separating Funnel with PTFE Stopcock

Manufactured from Borosilicate Glass and ISO 4800 Certified. Fisherbrand™ Glass Conical Separating Funnel with PTFE Stopcock is supplied with a glass and plastic stopper. Available with interchangeable glass or PTFE stopcock.

Brand:  Fisherbrand™ FB51743

111.22 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.


 View more versions of this product

Product Code. 11592782

  • 239.00 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

Manufactured from Borosilicate Glass and ISO 4800 Certified. Fisherbrand™ Glass Conical Separating Funnel with PTFE Stopcock is supplied with a glass and plastic stopper. Available with interchangeable glass or PTFE stopcock.

  • Manufactured from Borosilicate Glass
  • ISO 4800 Certified
  • Supplied with a glass and plastic stopper
  • Available with PTFE stopcock
TRUSTED_SUSTAINABILITY
Specifications

Specifications

500 mL
ISO 4800
PTFE
24/29
Borosilicate Glass
1PK
Glass, Plastic
Separatory Funnel
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ Glass Conical Separating Funnel with PTFE Stopcock > Capacity: 500mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.