Learn More
Hirschmann™ Glass covers
Remove stickiness and minimize dust. Hirschmann™ Glass covers are perfect for protecting and preserving microscopy slides.
Brand:  Hirschmann™ 8000108
 View more versions of this product
Product Code. 13148159
  • 47.05 € / Pack of 1000
Estimated Shipment: 26-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • LOT number on packaging
  • CE pursuant to IVD 98/79
  • DIN 58 884
Specifications
Specifications
#1 (0.13 to 0.16 mm)
21 x 26 mm