Learn More
Hirschmann™ Glass Coverslips for Haemacytometer
Cover glasses for Haemacytometer with smooth surface grinding, without CE mark.
Brand:  Hirschmann™ 8000125
 View more versions of this product
Product Code. 10161671
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Surface ground and polished surface
  • LOT number on the packaging
  • Labelled with: usable until, CE pursuant to IVD 98/79 EC According to the German Calibration Regulation (Eichordnung), only officially calibrated haemocytometer glass covers may be placed on the market and used in the FRG
Specifications
Specifications
Hemacytometer Coverslip
0.4 mm
Haemocytometer
Glass
22 x 22 mm