Learn More
Hanna Instruments™ Glass Cuvette Sets for Checker HC Colorimeters and Benchtop Photometers
Replacement optically clear glass cuvettes that are supplied with the Checker HC colorimeters and HI 83 series photometers and turbidimeters.
Brand:  Hanna Instruments™ HI731321
Product Code. 10751571
  • 54.35 € / Pack of 4
Estimated Shipment: 10-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For use with HI 83 series photometers and turbidimeters
  • Replacement glass cuvettes used with the HI7XX Checker HC colorimeters and the HI832XX benchtop photometers
  • Each cuvette has a 10 ml level mark to measure the appropriate volume
  • Index mark for proper alignment in the photometer
  • 10 ml level mark for adding correct volume
  • Optically clear glass
Specifications
Specifications
HI 83 Series Photometers and Turbidimeters