Learn More
Fisherbrand™ Glass-Encased Stir Bar
Use with very abrasive media that may erode PTFE. The glass-encased magnetic stir bar is cylindrical for a wide variety of uses.
Brand:  Fisherbrand™ 001.1206
 View more versions of this product
Product Code. 11537792
  • 25.70 € / Pack of 5
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Stable, general-purpose magnetic stir bar
  • Use with very abrasive media that may erode PTFE
  • Autoclavable
Specifications
Specifications
Autoclavable
Clear
Containers & Vessels / Stirrer Hot Plates
6 mm
Cylinder
Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
Glass encapsulated Alnico V magnet core
5 mm
5/Pk.