Learn More
Brady™ Miniature Circuit Breaker Lockouts - Pin-Out Wide
A safe and effective method for locking out European Circuit Breakers.
Brand:  Brady™ 090851
Product Code. 15613517
  • 123.00 € / Pack of 6
Estimated Shipment: 13-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Requires no tools to install
  • Lockout easily installs with the aid of a push button
  • Tie-bar style lockout has a thumb wheel for quick application
Specifications
Specifications
Circuit Breaker Lockout
Red/Black
UL 94 HB
Fiberglass-reinforced Plastic
ELEC. CIRCUIT BREAKER L/O EURO POW PK6
Fiberglass-reinforced Plastic
Miniature Circuit Breaker Lockouts - Pin-Out Wide
6 Pk.