Learn More
Macherey-Nagel™ Glass Flash Chromatography Column
293.00 € - 391.00 €
Specifications
Material Glass
Type Flash Column
Includes with Adapter and PTFE Tap
Column Type Flash Chromatography Glass Column
For Use With (Application) Suited The Separation of Compounds up to Gram Levels No Expensive Equipment Required
View More Specs
Products 8
Product Code Brand Length (Metric) Style Price Quantity & Availability  
Product Code Brand Length (Metric) Style Price Quantity & Availability  
10440132
View Documents
Macherey-Nagel™
727400
200 mm I.D.: 20 mm
293.00 €
Each
Estimated Shipment: 13-12-2024
to see stock.
Add to basket
10073131
View Documents
Macherey-Nagel™
727401
400 mm I.D.: 20 mm
302.00 €
Each
Estimated Shipment: 13-12-2024
to see stock.
Add to basket
10659771
View Documents
Macherey-Nagel™
727403
400 mm I.D.: 25 mm
313.00 €
Each
Estimated Shipment: 13-12-2024
to see stock.
Add to basket
10164801
View Documents
Macherey-Nagel™
727404
300 mm I.D.: 30 mm
321.00 €
Each
Estimated Shipment: 13-12-2024
to see stock.
Add to basket
10450132
View Documents
Macherey-Nagel™
727402
200 mm I.D.: 25 mm
302.00 €
Each
Estimated Shipment: 13-12-2024
to see stock.
Add to basket
10093141
View Documents
Macherey-Nagel™
727405
400 mm I.D.: 30 mm
330.00 €
Each
Estimated Shipment: 13-12-2024
to see stock.
Add to basket
10690152
View Documents
Macherey-Nagel™
727406
300 mm I.D.: 40 mm
341.00 €
Each
Estimated Shipment: 13-12-2024
to see stock.
Add to basket
10246281
View Documents
Macherey-Nagel™
727407
450 mm I.D.: 40 mm
391.00 €
Each
Estimated Shipment: 13-12-2024
to see stock.
Add to basket
Specifications
Glass
with Adapter and PTFE Tap
Suited The Separation of Compounds up to Gram Levels No Expensive Equipment Required
Flash Column
Flash Chromatography Glass Column