Learn More
Scientific Glass Labs™ Glass Gas Jars
Borosilicate glass construction. Ground flat rim provides seal for lid.
Brand:  Scientific Glass Labs™ 2704/50210
 View more versions of this product
Product Code. 12965931
  • 10.75 € / Each
Estimated Shipment: 09-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Borosilicate glass construction
  • Clear
  • Ground flat rim to seal with a flat lid
  • Lid not included
Specifications
Specifications
Glass
50 mm
1 Each
Gas Jar
210 mm