Learn More
Hirschmann™ Class AS Graduated Pipettes, Blue Graduation
Provides volumetric accuracy for your pipetting needs. Hirschmann Instruments™ Class AS Graduated Pipettes, Blue Graduation come with a cotton plug end as standard.
Brand:  Hirschmann™ 1101141
 View more versions of this product
Product Code. 15231209
  • 84.30 € / Pack of 12
Estimated Shipment: 23-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Acc. to DIN EN ISO 835
  • Main point ring graduation in blue
  • Graduated to tip, zero on top, type 3
  • Conformity certified, with dated batch identification
Specifications
Specifications
Yes
AS
Pipette filler works via a series of Glass ball valves for air release, suction and delivery
2 mL
Pipettes up to 1000 mL
DIN EN ISO 835
Natural Rubber
Blue
Pipette Filler