Learn More
Macherey-Nagel™ Glass Laboratory Sprayer
Used as an accessory in thin layer chromatography applications. Macherey-Nagel™ Glass Laboratory Sprayer includes a 100mL, clear glass conical flask with a sprayer-stopper assembly and rubber bulb/tubing.
Brand:  Macherey-Nagel™ 814101
140.93 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 10206621
  • 169.00 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Specifications
100 mL
Glass, Rubber
Laboratory Sprayer
Rubber bulb/tubing
Clear-Red
1 Ea.
TLC
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Macherey-Nagel™ Glass Laboratory Sprayer > Includes: Rubber bulb