Learn More
Hirschmann™ Class AS Schellbach Blue Graduated Micro Burettes
With Schellbach stripes for enhanced legibility.
Brand:  Hirschmann™ 3300252
 View more versions of this product
Product Code. 11732392
  • 455.00 € / Pack of 2
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Class AS
  • Manufactured from DURAN™ borosilicate glass for resistance to chemicals and thermal shock
  • Blue graduation marks
  • Certified conformity, with dated batch identification
  • Straight stopcock with PTFE key
  • Stopcock in filling tube with PTFE key
  • Main point ring graduation
  • Calibrated for EX
  • Acc. to DIN EN ISO 385
Specifications
Specifications
5 mL
0.02 mL
PTFE
Straight
±0.01 mL
DIN EN ISO 385
Glass
Schellbach Stripes
Blue
PTFE
Class AS
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Product Certifications
Special Offers
Special Offers
Product Content Correction
Product Title
Hirschmann™ Class AS Schellbach Blue Graduated Micro Burettes > Capacity: 5 mL