Learn More
Hirschmann™ minicaps™ Micro Capillary Pipettes, Disposable
Facilitates error-free capillary filling in the laboratory
Brand:  Hirschmann™ 9001133
 View more versions of this product
Product Code. 10267980
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Not heparinized
  • End-to-end minicaps
  • Acc. to ISO 9000, CE acc. IVD 98/79 EC
  • Color-coded vial cap with capillary dispenser
  • Conformity certified, with batch number
Specifications
Specifications
coefficent of variaton: 1%, accuracy: 0.5%
Disposable micro capillary pipettes, not heparinized, end-to-end; includes Pipette bulb (per 1000 pcs.)
10 x 100/Pack
Minicap Capillary Pipette
ISO 7550, CE acc. IVD 98/79 EG, conformity certified
Borosilicate Glass
3.33 μL
Single-use