Learn More
Hirschmann™ minicaps™ Micro Capillary Pipettes, Disposable
Facilitates error-free capillary filling in the laboratory
Brand:  Hirschmann™ 9000130
 View more versions of this product
Product Code. 10666751
  • 118.00 € / Pack of 1000
Estimated Shipment: 03-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Not heparinized
  • End-to-end, with minicaps
  • Acc. to ISO 9000, CE acc. IVD 98/79 EC
  • Color-coded vial cap with capillary dispenser
  • Conformity certified, with batch number
Specifications
Specifications
0.5%
Glass
30 to 32 mm
Single-use
ISO 7550, CE, IVD 98/79 EC
30 μL
Minicap Capillary Pipette