missing translation for 'onlineSavingsMsg'
Learn More

Hirschmann™ minicaps™ Micro Capillary Pipettes, Disposable

Facilitates error-free capillary filling in the laboratory

Brand:  Hirschmann™ 9000140

 View more versions of this product

Product Code. 11732293

  • 118.00 € / Pack of 1000
Estimated Shipment: 24-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Not heparinized
  • End-to-end minicaps
  • Acc. to ISO 9000, CE acc. IVD 98/79 EC
  • Color-coded vial cap with capillary dispenser
  • Conformity certified, with batch number
Specifications

Specifications

coefficent of variaton: 1%, accuracy: 0.5%
Disposable micro capillary pipettes, not heparinized, end-to-end; includes Pipette bulb (per 1000 pcs.)
10 x 100/Pack
Minicap Capillary Pipette
ISO 7550, CE acc. IVD 98/79 EG, conformity certified
Borosilicate Glass
40 μL
Single-use
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.