missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Soda Lime Glass Pasteur Pipette

Soda Lime Glass Pasteur Pipette is intended for single use in microbiological and serological laboratories.

Brand:  Fisherbrand™ M4150NO250FS4

39.89 EUR valid until 2025-02-28
Use promo code "24154" to get your promotional price.


 View more versions of this product

Product Code. 11546963

  • 93.60 € / Pack of 1000
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

  • Pasteur pipettes made of soda-lime glass are a good alternative to plastic pipettes
  • Complies with ISO 7712
  • Pasteur pipettes with short or long extended, fine tip (150 mm, 230 mm and 270 mm)
  • Pasteur pipettes with and without cotton plug
  • The filling volume is approx. 2 mL
  • Ideal for use with Fisherbrand Pipettors
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Not Autoclavable
Single Use in Microbiological and Serological Laboratory Applications, Pipetting Liquids, Dropwise Dosing of Liquids and Reagents, Distribution of Heated Liquid Quantities
Standard Tip
2 mL
Unplugged Pasteur Pipet
Fisherbrand Pipetters
Soda Lime Glass
1000 Pk.
0.07 oz.
150 mm
Single-use
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.